man

Test

CAS RN 138774-94-4

OHONHOO

CAS RN 138774-94-4

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(naphthalen-2-yl)propanoate
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(naphthalen-2-yl)propanoate
Molecular Weight (g/mol): 436.49
Molecular Formula: C28H22NO4
InChi Key: JYUTZJVERLGMQZ-AREMUKBSSA-M
SMILES: [O-]C(=O)[C@@H](CC1=CC=C2C=CC=CC2=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)