man

Test

CAS RN 138847-85-5

OONO

CAS RN 138847-85-5

IUPAC Name: 2-(morpholin-4-yl)ethyl 1-phenylcyclohexane-1-carboxylate
Synonyms: 2-(morpholin-4-yl)ethyl 1-phenylcyclohexane-1-carboxylate
Molecular Weight (g/mol): 317.43
Molecular Formula: C19H27NO3
InChi Key: RQHKZUBCUZVZEF-UHFFFAOYSA-N
SMILES: O=C(OCCN1CCOCC1)C1(CCCCC1)C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results