man

Test

CAS RN 13889-98-0

H3CONNH2

CAS RN 13889-98-0

IUPAC Name: 4-acetylpiperazin-1-ium
Synonyms: 4-acetylpiperazin-1-ium
Molecular Weight (g/mol): 129.18
Molecular Formula: C6H13N2O
InChi Key: PKDPUENCROCRCH-UHFFFAOYSA-O
SMILES: CC(=O)N1CC[NH2+]CC1
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (1)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (6)
Physical Form 
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (1)
  • (3)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)