CAS RN 13893-34-0
CAS RN 13893-34-0
IUPAC Name:
octadecane-1-sulfonic acid sodium
Synonyms:
octadecane-1-sulfonic acid sodium
Molecular Weight (g/mol):
357.55
Molecular Formula:
C18H38NaO3S
InChi Key:
NIMUYFTYMXTPAV-UHFFFAOYSA-N
SMILES:
[Na].CCCCCCCCCCCCCCCCCCS(O)(=O)=O
1
–
1
of
1
results
1
1-Octadecanesulfonic acid, sodium salt, 90%, Thermo Scientific Chemicals
CAS: 13893-34-0 Molecular Formula: C18H37NaO3S Molecular Weight (g/mol): 356.54 MDL Number: MFCD00051526 InChI Key: KBAFDSIZQYCDPK-UHFFFAOYSA-M Synonym: sodium 1-octadecanesulfonate,sodium octadecane-1-sulfonate,sodium stearylsulfonate,sodium octadecanesulfonate,1-octadecanesulfonic acid sodium salt,sodium octadecane-1-sulphonate,1-octadecanesulfonic acid, sodium salt,sodium sterylsulfonate,sodium oxadecanesulfonate,sodium n-octadecylsulfonate PubChem CID: 23668003 IUPAC Name: sodium;octadecane-1-sulfonate SMILES: CCCCCCCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]