man

Test

CAS RN 1390-65-4

CH3HOOHOOOHOHOOHOHOHHOOHO

CAS RN 1390-65-4

IUPAC Name: 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
Synonyms: carmine cochineal
Molecular Weight (g/mol): 492.39
Molecular Formula: C22H20O13
InChi Key: DGQLVPJVXFOQEV-UHFFFAOYNA-N
SMILES: CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
Color 
  • (2)
  • (3)
  • (2)
  • (1)
Quantity 
  • (2)
  • (3)
  • (3)
Grade 
  • (5)
Percent Purity 
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (5)
  • (3)