man

Test

CAS RN 13963-58-1

KKKCoCNCNCNCNCNCN

CAS RN 13963-58-1

IUPAC Name: cobalt(3+) tripotassium hexakis(iminomethanide)
Synonyms: cobalt(3+) tripotassium hexakis(cyanide)
Molecular Weight (g/mol): 332.34
Molecular Formula: C6CoK3N6
InChi Key: LGRDAQPMSDIUQJ-UHFFFAOYSA-N
SMILES: [K+].[K+].[K+].[Co+3].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)