man

Test

CAS RN 14050-01-2

NOONOOCu

CAS RN 14050-01-2

IUPAC Name: copper(2+) bis(pyridine-2-carboxylate)
Synonyms: copper(2+) dipicolinate
Molecular Weight (g/mol): 307.75
Molecular Formula: C12H8CuN2O4
InChi Key: GNWVUSJJFONIKF-UHFFFAOYSA-L
SMILES: [Cu++].[O-]C(=O)C1=CC=CC=N1.[O-]C(=O)C1=CC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results