man

Test

CAS RN 14059-33-7

OOOOVBi

CAS RN 14059-33-7

IUPAC Name: bismuth(3+) vanadium tetraoxidandiide
Synonyms: bismuth vanadate
Molecular Weight (g/mol): 323.92
Molecular Formula: BiO4V
InChi Key: HUUOUJVWIOKBMD-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[V].[Bi+3]

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results