CAS RN 141-95-7
CAS RN 141-95-7
IUPAC Name:
disodium propanedioate
Synonyms:
sodium malonate propanedioic acid, sodium salt (1:2) sodium malonate dibasic propanedioic acid, disodium salt malonic acid, sodium salt malonic acid, disodium salt malonic acid disodium salt
Molecular Weight (g/mol):
148.03
Molecular Formula:
C3H2Na2O4
InChi Key:
PRWXGRGLHYDWPS-UHFFFAOYSA-L
SMILES:
[Na+].[Na+].[O-]C(=O)CC([O-])=O
1
–
1
of
1
results
1
Malonic acid disodium salt, 99%, Thermo Scientific Chemicals
CAS: 141-95-7 Molecular Formula: C3H2Na2O4 Molecular Weight (g/mol): 148.025 MDL Number: MFCD00002708 InChI Key: PRWXGRGLHYDWPS-UHFFFAOYSA-L Synonym: sodium malonate,disodium malonate,propanedioic acid, disodium salt,malonic acid disodium salt,disodium propanedioate,malonic acid, sodium salt,unii-9qwe64m39h,propanedioic acid, sodium salt 1:2,malonic acid, disodium salt,dicarboxymethane disodium salt PubChem CID: 8865 ChEBI: CHEBI:62983 IUPAC Name: disodium;propanedioate SMILES: C(C(=O)[O-])C(=O)[O-].[Na+].[Na+]