man

Test

CAS RN 1412905-42-0

BOOH3CCH3H3CH3COH2NF

CAS RN 1412905-42-0

IUPAC Name: 4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Synonyms: 4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Molecular Weight (g/mol): 265.09
Molecular Formula: C13H17BFNO3
InChi Key: RBEAFMIZDXOHAT-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(F)C=CC(=C1)C(N)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results