man

Test

CAS RN 141699-55-0

CH3H3CCH3OONOH

CAS RN 141699-55-0

IUPAC Name: tert-butyl 3-hydroxyazetidine-1-carboxylate
Synonyms: tert-butyl 3-hydroxyazetidine-1-carboxylate
Molecular Weight (g/mol): 173.21
Molecular Formula: C8H15NO3
InChi Key: XRRXRQJQQKMFBC-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CC(O)C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)