CAS RN 14175-03-2
CAS RN 14175-03-2
IUPAC Name:
tris(oxalic acid) decahydrate disamarium
Synonyms:
tris(oxalic acid) decahydrate disamarium
Molecular Weight (g/mol):
750.97
Molecular Formula:
C6H26O22Sm2
InChi Key:
IKPDQYODFRQYAD-UHFFFAOYSA-N
SMILES:
O.O.O.O.O.O.O.O.O.O.[Sm].[Sm].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O
1
–
2
of
2
results
1
Samarium(III) oxalate hydrate, REacton™, 99.97% (REO), Thermo Scientific™
CAS: 14175-03-2 Molecular Formula: C6H26O22Sm2 Molecular Weight (g/mol): 750.97 MDL Number: MFCD00150489 InChI Key: IKPDQYODFRQYAD-UHFFFAOYSA-N Synonym: samarium iii oxalate hydrate 2:3:10,ethanedioic acid,samarium 3+ salt, hydrate 3:2:10 PubChem CID: 121488066 SMILES: O.O.O.O.O.O.O.O.O.O.[Sm].[Sm].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O
2
Samarium(III) oxalate decahydrate, 99.9% (REO), Thermo Scientific™
CAS: 14175-03-2 Molecular Formula: C6H26O22Sm2 Molecular Weight (g/mol): 750.97 MDL Number: MFCD00150489 InChI Key: IKPDQYODFRQYAD-UHFFFAOYSA-N Synonym: samarium iii oxalate hydrate 2:3:10,ethanedioic acid,samarium 3+ salt, hydrate 3:2:10 PubChem CID: 121488066 IUPAC Name: oxalic acid;samarium;decahydrate SMILES: O.O.O.O.O.O.O.O.O.O.[Sm].[Sm].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O