man

Test

CAS RN 14284-89-0

OOOOOOH3CCH3CH3H3CH3CH3CMn

CAS RN 14284-89-0

IUPAC Name: manganese(3+) 4-oxopent-2-en-2-olate
Synonyms: manganese(3+) 4-oxopent-2-en-2-olate
Molecular Weight (g/mol): 154.05
Molecular Formula: C5H7MnO2
InChi Key: BCOBLSSFEZDWKH-UHFFFAOYSA-M
SMILES: [Mn+3].CC([O-])=CC(C)=O
Physical Form 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)