CAS RN 143313-51-3
CAS RN 143313-51-3
IUPAC Name:
(3S)-3-{[(2S)-1-[(2S)-2-[(2S)-2-acetamido-3-(4-hydroxyphenyl)propanamido]-3-methylbutanamido]-1-oxopropan-2-yl]amino}-4-oxobutanoic acid
Synonyms:
(3S)-3-{[(2S)-1-[(2S)-2-[(2S)-2-acetamido-3-(4-hydroxyphenyl)propanamido]-3-methylbutanamido]-1-oxopropan-2-yl]amino}-4-oxobutanoic acid
Molecular Weight (g/mol):
492.53
Molecular Formula:
C23H32N4O8
InChi Key:
KQCBPHUOVBKBPQ-SJVNDZIOSA-N
SMILES:
CC(C)[C@H](NC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O)C(=O)NC(=O)[C@H](C)N[C@@H](CC(O)=O)C=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Caspase-1 Inhibitor I
CAS: 143313-51-3 Molecular Formula: C23H32N4O8 Synonym: Ac-YVAD-CHO; Ac-Tyr-Val-Ala-Asp-CHO