CAS RN 143824-78-6
CAS RN 143824-78-6
IUPAC Name:
3-{1-[(tert-butoxy)carbonyl]-1H-indol-3-yl}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms:
3-[1-(tert-butoxycarbonyl)indol-3-yl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol):
526.59
Molecular Formula:
C31H30N2O6
InChi Key:
ADOHASQZJSJZBT-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)N1C=C(CC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C2=CC=CC=C12
1
–
1
of
1
results
1
1-Boc-N-Fmoc-L-tryptophan, 97%, Thermo Scientific Chemicals
CAS: 143824-78-6 Molecular Formula: C31H30N2O6 Molecular Weight (g/mol): 526.59 MDL Number: MFCD00153366 InChI Key: ADOHASQZJSJZBT-SANMLTNESA-N Synonym: fmoc-trp boc-oh,fmoc-l-trp boc-oh,nalpha-9h-fluoren-9-ylmethoxy carbonyl-n1-tert-butoxycarbonyl-l-tryptophan,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1-tert-butoxycarbonyl-1h-indol-3-yl propanoic acid,2s-3-1-tert-butoxycarbonyl indol-3-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,fmoc-l-trp boc,pubchem10048,1-boc-n-fmoc-l-tryptophan,ksc910o8t,n1-boc-nalpha-fmoc-l-tryptophan PubChem CID: 9849766 SMILES: CC(C)(C)OC(=O)N1C=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12