man

Test

CAS RN 14389-86-7

OOO

CAS RN 14389-86-7

IUPAC Name: 2-(benzyloxy)benzoate
Synonyms: 2-(benzyloxy)benzoate
Molecular Weight (g/mol): 227.24
Molecular Formula: C14H11O3
InChi Key: GMOYUTKNPLBTMT-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC=C1OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results