man

Test

CAS RN 1452-94-4

H3COONCl

CAS RN 1452-94-4

IUPAC Name: ethyl 2-chloropyridine-3-carboxylate
Synonyms: ethyl 2-chloropyridine-3-carboxylate
Molecular Weight (g/mol): 185.61
Molecular Formula: C8H8ClNO2
InChi Key: PMIMPBYTPPRBGD-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CN=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results