man

Test

CAS RN 14532-05-9

CH3CH3OCH3CH3OCH3CH3OGd

CAS RN 14532-05-9

IUPAC Name: gadolinium(3+) tris(propan-2-olate)
Synonyms: gadolinium(3+) tris(propan-2-olate)
Molecular Weight (g/mol): 334.51
Molecular Formula: C9H21GdO3
InChi Key: VJLSFXQJAXVOEQ-UHFFFAOYSA-N
SMILES: [Gd+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)