man

Test

CAS RN 1457-46-1

OSSN

CAS RN 1457-46-1

IUPAC Name: 3-phenyl-2-sulfanylidene-1,3-thiazolidin-4-one
Synonyms: 3-phenyl-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol): 209.28
Molecular Formula: C9H7NOS2
InChi Key: DVRWEKGUWZINTQ-UHFFFAOYSA-N
SMILES: O=C1CSC(=S)N1C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results