man

Test

CAS RN 14694-95-2

RhPPPClPhPhPhPhPhPhPhPhPh

CAS RN 14694-95-2

IUPAC Name: tris(triphenylphosphane) hydrochloridyl rhodium
Synonyms: wilkinson's catalyst
Molecular Weight (g/mol): 925.23
Molecular Formula: C54H45ClP3Rh
InChi Key: TZKOFQQLYGJWKC-UHFFFAOYSA-N
SMILES: [Cl].[Rh].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Grade 
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)