man

Test

CAS RN 14752-66-0

OSOClNa

CAS RN 14752-66-0

IUPAC Name: 4-chlorobenzene-1-sulfinate
Synonyms: 4-chlorobenzenesulfinate
Molecular Weight (g/mol): 175.61
Molecular Formula: C6H4ClO2S
InChi Key: AOQYAMDZQAEDLO-UHFFFAOYSA-M
SMILES: [O-]S(=O)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)