man

Test

CAS RN 1476-23-9

CH2NCO

CAS RN 1476-23-9

IUPAC Name: 3-isocyanatoprop-1-ene
Synonyms: 1-propene, 3-isocyanato-
Molecular Weight (g/mol): 83.09
Molecular Formula: C4H5NO
InChi Key: HXBPYFMVGFDZFT-UHFFFAOYSA-N
SMILES: C=CCN=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results