CAS RN 148-79-8
CAS RN 148-79-8
IUPAC Name:
2-(1,3-thiazol-4-yl)-1H-1,3-benzodiazole
Synonyms:
testo triasox thibenzol top form wormer syntol M100 lombristop epa pesticide chemical code 060101 thiabendazole (product) tubazole ormogal
Molecular Weight (g/mol):
201.25
Molecular Formula:
C10H7N3S
InChi Key:
WJCNZQLZVWNLKY-UHFFFAOYSA-N
SMILES:
N1C2=CC=CC=C2N=C1C1=CSC=N1
1
–
8
of
8
results