man

Test

CAS RN 148490-97-5

H3COIOOCH3

CAS RN 148490-97-5

IUPAC Name: methyl 4-iodo-2-methoxybenzoate
Synonyms: methyl 4-iodo-2-methoxybenzoate
Molecular Weight (g/mol): 292.07
Molecular Formula: C9H9IO3
InChi Key: MCNOTXROWOGSGU-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(OC)C=C(I)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results