man

Test

CAS RN 14855-76-6

ClBrH3CNH3CCH3CNCH3H3CNCH3CH3

CAS RN 14855-76-6

IUPAC Name: bis[4-(dimethylamino)phenyl][4-(ethyldimethylazaniumyl)phenyl]methylium bromide chloride
Synonyms: ethyl green
Molecular Weight (g/mol): 516.95
Molecular Formula: C27H35BrClN3
InChi Key: IDAQSADEMXDTKN-UHFFFAOYSA-L
SMILES: [Cl-].[Br-].CC[N+](C)(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results