CAS RN 150-84-5
CAS RN 150-84-5
IUPAC Name:
3,7-dimethyloct-6-en-1-yl acetate
Synonyms:
citronellyl acetate (S)-isomer citronellyl acetate citronellyl ethanoate (s)-citronellyl acetate (s)-isomer citronellyl acetate natural rhodinol, acetylated acetylated natural rhodinol acetic acid, citronellyl ester citronellol acetate ctronellyl acetate (natural)
Molecular Weight (g/mol):
198.31
Molecular Formula:
C12H22O2
InChi Key:
JOZKFWLRHCDGJA-UHFFFAOYNA-N
SMILES:
CC(CCOC(C)=O)CCC=C(C)C
1
–
1
of
1
results
1
Citronellyl acetate, 96%, Thermo Scientific Chemicals
CAS: 150-84-5 Molecular Formula: C12H22O2 Molecular Weight (g/mol): 198.306 MDL Number: MFCD00015039 InChI Key: JOZKFWLRHCDGJA-UHFFFAOYSA-N Synonym: citronellyl acetate,citronellol acetate,3,7-dimethyl-6-octen-1-yl acetate,3,7-dimethyloct-6-en-1-yl acetate,1-acetoxy-3,7-dimethyloct-6-ene,acetic acid, citronellyl ester,natural rhodinol, acetylated,citronellyl ethanoate,b-citronellyl acetate,6-octen-1-ol, 3,7-dimethyl-, acetate PubChem CID: 9017 ChEBI: CHEBI:70478 IUPAC Name: 3,7-dimethyloct-6-enyl acetate SMILES: CC(CCC=C(C)C)CCOC(=O)C