man

Test

CAS RN 15067-26-2

2H2H2H2H2H2H2H2H2H2H

CAS RN 15067-26-2

IUPAC Name: 1,2-dihydro(²H₁₀)acenaphthylene
Synonyms: (²H₁₀)acenaphthylene
Molecular Weight (g/mol): 164.27
Molecular Formula: C12H10
InChi Key: CWRYPZZKDGJXCA-WHUVPORUSA-N
SMILES: [2H]C1=C([2H])C2=C([2H])C([2H])=C([2H])C3=C2C(=C1[2H])C([2H])([2H])C3([2H])[2H]
For Use With (Equipment) 
  • (2)
Quantity 
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results