man

Test

CAS RN 15104-46-8

CH3OPOOCH3POOH3COH3C

CAS RN 15104-46-8

IUPAC Name: dimethyl [2-(dimethoxyphosphoryl)phenyl]phosphonate
Synonyms: dimethyl 2-(dimethoxyphosphoryl)phenylphosphonate
Molecular Weight (g/mol): 294.18
Molecular Formula: C10H16O6P2
InChi Key: TUKTVDDATWNXSN-UHFFFAOYSA-N
SMILES: COP(=O)(OC)C1=CC=CC=C1P(=O)(OC)OC
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results