man

Test

CAS RN 1513-69-5

NH2NOHNFFF

CAS RN 1513-69-5

IUPAC Name: 2-amino-6-(trifluoromethyl)-1,4-dihydropyrimidin-4-one
Synonyms: 2-amino-6-(trifluoromethyl)-1H-pyrimidin-4-one
Molecular Weight (g/mol): 179.1
Molecular Formula: C5H4F3N3O
InChi Key: ZEPSVMLZBXDPGU-UHFFFAOYSA-N
SMILES: NC1=NC(=O)C=C(N1)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results