man

Test

CAS RN 1524-40-9

NH2SOOF

CAS RN 1524-40-9

IUPAC Name: 3-fluorobenzene-1-sulfonamide
Synonyms: 3-fluorobenzenesulfonamide
Molecular Weight (g/mol): 175.18
Molecular Formula: C6H6FNO2S
InChi Key: CRINBBOGNYCAOV-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=CC(F)=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)