man

Test

CAS RN 15274-78-9

H2OH2OH2OH2OOSOOOOSOOOOSOOORhRh

CAS RN 15274-78-9

IUPAC Name: dirhodium(3+) tetrahydrate trisulfate
Synonyms: dirhodium(3+) tetrahydrate trisulfate
Molecular Weight (g/mol): 566.04
Molecular Formula: H8O16Rh2S3
InChi Key: JJITWKYISWZJBI-UHFFFAOYSA-H
SMILES: O.O.O.O.[Rh+3].[Rh+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Grade 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)