man

Test

CAS RN 15283-45-1

OSOSOOSOSONaNaNaAu

CAS RN 15283-45-1

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 492.19
Molecular Formula: AuH2Na3O6S4
InChi Key: UCGZDNYYMDPSRK-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[Na+].[Au].OS([S-])(=O)=O.OS([S-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results