man

Test

CAS RN 1535-73-5

NH2OFFF

CAS RN 1535-73-5

IUPAC Name: 3-(trifluoromethoxy)aniline
Synonyms: 3-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 177.13
Molecular Formula: C7H6F3NO
InChi Key: SADHVOSOZBAAGL-UHFFFAOYSA-N
SMILES: NC1=CC=CC(OC(F)(F)F)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)