CAS RN 155213-67-5
CAS RN 155213-67-5
IUPAC Name:
(1,3-thiazol-5-yl)methyl N-[(2S,3S,5S)-3-hydroxy-5-[(2S)-3-methyl-2-{[methyl({[2-(propan-2-yl)-1,3-thiazol-4-yl]methyl})carbamoyl]amino}butanamido]-1,6-diphenylhexan-2-yl]carbamate
Synonyms:
ritonavir 利托那韦 RTV norvir norvir sec
Molecular Weight (g/mol):
720.95
Molecular Formula:
C37H48N6O5S2
InChi Key:
NCDNCNXCDXHOMX-XGKFQTDJSA-N
SMILES:
CC(C)[C@H](NC(=O)N(C)CC1=CSC(=N1)C(C)C)C(=O)N[C@H](C[C@H](O)[C@H](CC1=CC=CC=C1)NC(=O)OCC1=CN=CS1)CC1=CC=CC=C1
1
–
3
of
3
results
1
Ritonavir, 98%, Thermo Scientific Chemicals
CAS: 155213-67-5 Molecular Formula: C37H48N6O5S2 Molecular Weight (g/mol): 720.94 InChI Key: NCDNCNXCDXHOMX-XGKFQTDJSA-N Synonym: ritonavir,norvir,abbott 84538,abbott-84538,ritonavir usan,norvir softgel,norvir sec,unii-o3j8g9o825,norvir tm,norvir tn PubChem CID: 392622 ChEBI: CHEBI:45409 IUPAC Name: 1,3-thiazol-5-ylmethyl N-[(2S,3S,5S)-3-hydroxy-5-[[(2S)-3-methyl-2-[[methyl-[(2-propan-2-yl-1,3-thiazol-4-yl)methyl]carbamoyl]amino]butanoyl]amino]-1,6-diphenylhexan-2-yl]carbamate SMILES: CC(C)C1=NC(=CS1)CN(C)C(=O)NC(C(C)C)C(=O)NC(CC2=CC=CC=C2)CC(C(CC3=CC=CC=C3)NC(=O)OCC4=CN=CS4)O
2
Tocris Bioscience™ Ritonavir
HIV-1 and HIV-2 protease inhibitor
3