man

Test

CAS RN 15586-77-3

OSiOOCsCs

CAS RN 15586-77-3

IUPAC Name: dicaesium(1+) oxosilanebis(olate)
Synonyms: dicaesium(1+) oxosilanebis(olate)
Molecular Weight (g/mol): 341.89
Molecular Formula: Cs2O3Si
InChi Key: MZRJYMUQLBFTMG-UHFFFAOYSA-N
SMILES: [Cs+].[Cs+].[O-][Si]([O-])=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results