man

Test

CAS RN 156478-71-6

CH3H3CCH3OONNOHO

CAS RN 156478-71-6

IUPAC Name: 2-{4-[(tert-butoxy)carbonyl]piperazin-1-yl}acetic acid
Synonyms: [4-(tert-butoxycarbonyl)piperazin-1-yl]acetic acid
Molecular Weight (g/mol): 244.29
Molecular Formula: C11H20N2O4
InChi Key: WZBHMXRBXXCEDD-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC(O)=O)CC1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results