man

Test

CAS RN 1567-75-5

CH3OH3C

CAS RN 1567-75-5

IUPAC Name: 1-(1-methylcyclopropyl)ethan-1-one
Synonyms: 1-(1-methylcyclopropyl)ethanone
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: OQBCJXUAQQMTRW-UHFFFAOYSA-N
SMILES: CC(=O)C1(C)CC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)