man

Test

CAS RN 15742-38-8

NH3NH3NH3NH3NH3ClIrClCl

CAS RN 15742-38-8

IUPAC Name: trichloroiridium pentaamine
Synonyms: trichloroiridium pentaamine
Molecular Weight (g/mol): 383.72
Molecular Formula: Cl3H15IrN5
InChi Key: DYGMZANLQHDDSH-UHFFFAOYSA-K
SMILES: N.N.N.N.N.Cl[Ir](Cl)Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results