man

Test

CAS RN 1575-37-7

NH2BrH2N

CAS RN 1575-37-7

IUPAC Name: 4-bromobenzene-1,2-diamine
Synonyms: 4-bromobenzene-1,2-diamine
Molecular Weight (g/mol): 187.04
Molecular Formula: C6H7BrN2
InChi Key: WIHHVKUARKTSBU-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results