CAS RN 15845-62-2
CAS RN 15845-62-2
IUPAC Name:
1-iodo-4-isocyanatobenzene
Synonyms:
1-iodo-4-isocyanatobenzene
Molecular Weight (g/mol):
245.02
Molecular Formula:
C7H4INO
InChi Key:
KVMKUTIUJGYHBA-UHFFFAOYSA-N
SMILES:
IC1=CC=C(C=C1)N=C=O
1
–
1
of
1
results
1
4-Iodophenyl isocyanate, 97%, Thermo Scientific Chemicals
CAS: 15845-62-2 Molecular Formula: C7H4INO Molecular Weight (g/mol): 245.019 MDL Number: MFCD00013875 InChI Key: KVMKUTIUJGYHBA-UHFFFAOYSA-N Synonym: 4-iodophenyl isocyanate,4-iodophenylisocyanate,1-iodo-4-isocyanato-benzene,p-iodophenylisocyanate,acmc-1bsbj,benzene,1-iodo-4-isocyanato PubChem CID: 3399787 IUPAC Name: 1-iodo-4-isocyanatobenzene SMILES: C1=CC(=CC=C1N=C=O)I