Learn More
0.0 NTU Ratio Turbidity Standard, Reagecon
Suspension of extremely accurate (± 1%) polymer microspheres with guaranteed shelf life stability even after opening the bottle.
Brand: Reagecon CRSR-0-500
Description
- Turbidity Reference Standard, Turbidity Reference Material.
- Designed and formulated to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards, primarily formazin-based standards.
- Certified
- NIST traceable
- Non-toxic, non-carcinogenic
- Highly accurate
- Stable
- EPA approved
Specifications
Non-Ratio Instrumentation | |
Accuracy ± 1% NTU | |
0.0 NTU ± 1% | |
500 mL |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
CRSR00524L1 | Certificate of Analysis | 23/12/2024 | CRSR-0-500 |
CRSR00523C1 | Certificate of Analysis | 28/06/2023 | CRSR-0-500 |
CRSR00522M1 | Certificate of Analysis | 08/03/2023 | CRSR-0-500 |
CRSR00522G1 | Certificate of Analysis | 25/08/2022 | CRSR-0-500 |
CRSR00521J1 | Certificate of Analysis | 04/05/2022 | CRSR-0-500 |