missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ 0.8μm Pore Cellulose Nitrate Membrane

Brand:  Sartorius™ 13004--47----ACN


Product Code. 10478742

  • 1670.00 SEK / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Black/White Grid
Cellulose Nitrate Membrane Filter
0.8 μm
Microbiological testing of beverages, yeasts and molds. For testing airborne yeasts and molds at places high humidity
100/Pack
47 mm
1.7bar
Cellulose Nitrate Membrane Filter
Cellulose Nitrate
200mL/min.
5.2L/min.
0.14 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • X100 FILTERS,MEMBRANE DISC STERILE GREY WITH D=47mm, grau/weiß,

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers