missing translation for 'onlineSavingsMsg'
Learn More
1,1,1-Trifluoroacetone cyanohydrin, 95%, Thermo Scientific Chemicals Product Code.: 11377696

1,1,1-Trifluoroacetone cyanohydrin, 95%, Thermo Scientific Chemicals

Product Code. 11377696
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11377696

Brand: Thermo Scientific Alfa Aesar L11318.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A chlorinated acetone derivative for organic synthesis and proteomics research

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A chlorinated acetone derivative for organic synthesis and proteomics research

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-08-0
Molecular Formula C4H4F3NO
Molecular Weight (g/mol) 139.077
MDL Number MFCD00040885
InChI Key XDCMNDCKYSQKAX-UHFFFAOYSA-N
Synonym 1,1,1-trifluoroacetone cyanohydrin, 2-cyano-1,1,1-trifluoro-2-propanol, 2-hydroxy-2-trifluoromethyl propionitrile, propanenitrile, 3,3,3-trifluoro-2-hydroxy-2-methyl, 2-trifluoromethyl lactonitrile, trifluoroacetone cyanohydrin, lactonitrile, 2-trifluoromethyl, 3-trifluoro-a-hydroxyisobutyronitrile, lactonitrile,3,3-trifluoro-2-methyl, 2-cyano-1,1,1-trifluoropropan-2-olShow MoreShow Less
PubChem CID 274400
IUPAC Name 3,3,3-trifluoro-2-hydroxy-2-methylpropanenitrile
SMILES CC(C#N)(C(F)(F)F)O

Specifications

Density 1.276
Boiling Point 68°C to 69°C (40 mmHg)
Flash Point 60°C (140°F)
Refractive Index 1.338
Quantity 5 g
UN Number UN2929
Beilstein 1758723
Sensitivity Moisture sensitive
Solubility Information Fully miscible in water.
Formula Weight 139.08
Percent Purity 95%
Chemical Name or Material 1,1,1-Trifluoroacetone cyanohydrin
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247374 Certificate TypeCertificate of Analysis Date17/07/2023 Catalog NumberL11318.06,L11318.03,L11318.14
Lot Number10191109 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL11318.06,L11318.14
Lot Number10158620 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL11318.06,L11318.14
Lot NumberG24R071 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL11318.06,L11318.14
Lot NumberH06S033 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL11318.06,L11318.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,1,1-Trifluoroacetone cyanohydrin, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.