missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1-Trifluoroacetone cyanohydrin, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
A chlorinated acetone derivative for organic synthesis and proteomics research
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A chlorinated acetone derivative for organic synthesis and proteomics research
Solubility
Fully miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
A chlorinated acetone derivative for organic synthesis and proteomics research
Solubility
Fully miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.

Specifications
Specifications
Density | 1.276 |
Boiling Point | 68°C to 69°C (40 mmHg) |
Flash Point | 60°C (140°F) |
Refractive Index | 1.338 |
Quantity | 1 g |
UN Number | UN2929 |
Beilstein | 1758723 |
Sensitivity | Moisture sensitive |
Solubility Information | Fully miscible in water. |
Formula Weight | 139.08 |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1,1,1-Trifluoroacetone cyanohydrin, 95%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction