missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3-Hexanetriol, 97%, Thermo Scientific™
2194.00 SEK
Chemical Identifiers
CAS | 90325-47-6 |
---|---|
Molecular Formula | C6H14O3 |
Molecular Weight (g/mol) | 134.18 |
InChI Key | XYXCXCJKZRDVPU-UHFFFAOYSA-N |
Synonym | 1,2,3-hexanetriol, rac-syn-hexane-1,2,3-triol, dsstox_cid_31384, dsstox_gsid_52884, 1,2,3-hexanetriol solution, 1,2,3-hexanetriol 1 m solution, 1,2,3-hexanetriol, bioxtra gc, additive screening solution 07/fluka kit no 78374Show More |
PubChem CID | 2724429 |
IUPAC Name | hexane-1,2,3-triol |
SMILES | CCCC(C(CO)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15790839
|
Thermo Scientific Acros
232510050 |
5 g | N/A | ||||||
Description
Chemical Identifiers
90325-47-6 | |
134.18 | |
1,2,3-hexanetriol, rac-syn-hexane-1,2,3-triol, dsstox_cid_31384, dsstox_gsid_52884, 1,2,3-hexanetriol solution, 1,2,3-hexanetriol 1 m solution, 1,2,3-hexanetriol, bioxtra gc, additive screening solution 07/fluka kit no 78374Show More | |
hexane-1,2,3-triol |
C6H14O3 | |
XYXCXCJKZRDVPU-UHFFFAOYSA-N | |
2724429 | |
CCCC(C(CO)O)O |
Specifications
90325-47-6 | |
Authentic | |
C6H14O3 | |
1,2,3-hexanetriol, rac-syn-hexane-1,2,3-triol, dsstox_cid_31384, dsstox_gsid_52884, 1,2,3-hexanetriol solution, 1,2,3-hexanetriol 1 m solution, 1,2,3-hexanetriol, bioxtra gc, additive screening solution 07/fluka kit no 78374 | |
XYXCXCJKZRDVPU-UHFFFAOYSA-N | |
hexane-1,2,3-triol | |
2724429 | |
97% |
67.0°C to 69.0°C | |
96% min. (GC) | |
CH3(CH2)2CHOHCHOHCH2OH | |
Solubility in water: miscible in water. | |
CCCC(C(CO)O)O | |
134.18 | |
134.18 | |
1,2,3-Hexanetriol |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1,2,3-Hexanetriol, 97%, Thermo Scientific™