missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane, 97+%, Thermo Scientific Chemicals

Product Code. 15475996
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is a DuPhos and BPE ligands which are highly efficient privileged ligands. It is a chiral phosphine ligand used in the asymmetric hydrogenation of allylic sulfoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a DuPhos and BPE ligands which are highly efficient privileged ligands. It is a chiral phosphine ligand used in the asymmetric hydrogenation of allylic sulfoxides.

Solubility
Sparingly Soluble (3.2E-5 g/L) (25°C).

Notes
Incompatible with strong oxidizing agents. Store under room temperature. Air sensitive.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 824395-67-7
Molecular Formula C34H36P2
Molecular Weight (g/mol) 506.61
MDL Number MFCD07369028
InChI Key VHHAZLMVLLIMHT-CUPIEXAXSA-N
Synonym +-1,2-bis 2s,5s-2,5-diphenylphospholano ethane, 1,2-bis 2s,5s-2,5-diphenylphospholan-1-yl ethane, 1,1'-ethylenebis 2alpha,5beta-diphenyltetrahydro-1h-phosphole, +-1,2-bis 2s,5s-2,5-diphenylphospholano ethane, kanata purity, 2s,5s-1-2-2s,5s-2,5-diphenylphospholan-1-yl ethyl-2,5-diphenylphospholane
PubChem CID 16218207
IUPAC Name (2S,5S)-1-[2-[(2S,5S)-2,5-diphenylphospholan-1-yl]ethyl]-2,5-diphenylphospholane
SMILES C1CC(P(C1C2=CC=CC=C2)CCP3C(CCC3C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6

Specifica

Melting Point 143°C to 147°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Sparingly Soluble (3.2E-5 g/L) (25°C).
Formula Weight 506.6
Percent Purity ≥97%
Chemical Name or Material 1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane, 97+%, Thermo Scientific Chemicals >

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato