missing translation for 'onlineSavingsMsg'
Learn More
1-(3,5-Dichlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™ Product Code.: 10069471

1-(3,5-Dichlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10069471
10 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 10069471

Brand: Thermo Scientific Maybridge SP01487EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 306936-60-7
Molecular Formula C13H12Cl2N2O2
Molecular Weight (g/mol) 299.151
MDL Number MFCD02677727
InChI Key CIAUICOJDLJNBL-UHFFFAOYSA-N
Synonym 1-3,5-dichlorophenyl-5-propyl-1h-pyrazole-4-carboxylic acid, 1-3,5-dichlorophenyl-5-propylpyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylicacid, 1-3,5-dichlorophenyl-5-propyl, maybridge3_006729, 4-carboxy-1-3,5-dichlorophenyl-5-propyl-1h-pyrazoleShow MoreShow Less
PubChem CID 2743017
IUPAC Name 1-(3,5-dichlorophenyl)-5-propylpyrazole-4-carboxylic acid
SMILES CCCC1=C(C=NN1C2=CC(=CC(=C2)Cl)Cl)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 299.16
Percent Purity 97%
Chemical Name or Material 1-(3, 5-Dichlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(3,5-Dichlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.