Learn More
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, 97%, Thermo Scientific Chemicals
614.00 SEK - 6900.00 SEK
Chemical Identifiers
CAS | 69304-37-6 |
---|---|
Molecular Formula | C12H28Cl2OSi2 |
Molecular Weight (g/mol) | 315.425 |
MDL Number | MFCD00009655 |
InChI Key | DDYAZDRFUVZBMM-UHFFFAOYSA-N |
Synonym | 1,3-dichloro-1,1,3,3-tetraisopropyldisiloxane, 1,3-dichlorotetraisopropyldisiloxane, 1,1,3,3-tetraisopropyl-1,3-dichlorodisiloxane, 1,3-dichloro-1,1,3,3-tetraisopropyl-disiloxane, disiloxane, 1,3-dichloro-1,1,3,3-tetrakis 1-methylethyl, 1,1,3,3-tetraisopropyl-1,3-dichlorosiloxane, chloro chlorodiisopropylsilyl oxy diisopropylsilane, chloro-chloro-di propan-2-yl silyl oxy-di propan-2-yl silane, tipdsicl2, tipdsi-cl2Show More |
PubChem CID | 172404 |
IUPAC Name | chloro-[chloro-di(propan-2-yl)silyl]oxy-di(propan-2-yl)silane |
SMILES | CC(C)[Si](C(C)C)(O[Si](C(C)C)(C(C)C)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307616
|
Thermo Scientific Alfa Aesar
L11171.03 |
1 g | N/A | ||||||
11317616
|
Thermo Scientific Alfa Aesar
L11171.06 |
5 g |
2055.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327616
|
Thermo Scientific Alfa Aesar
L11171.14 |
25 g |
6900.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane is a silylating reagent used for simultaneous protection of 3'- and 5'-hydroxy functions of ribonucleosides. It is used in the analysis of Watson-Crick and Hoogstein base pairing in nucleotides. It is also employed in the formation of ribavirin chemical delivery systems. Further, it serves as a protecting group reagent for open-chain polyhydroxy compounds. In addition to this, it is involved in the preparation of cyclic bridged peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane is a silylating reagent used for simultaneous protection of 3′- and 5′-hydroxy functions of ribonucleosides. It is used in the analysis of Watson-Crick and Hoogstein base pairing in nucleotides. It is also employed in the formation of ribavirin chemical delivery systems. Further, it serves as a protecting group reagent for open-chain polyhydroxy compounds. In addition to this, it is involved in the preparation of cyclic bridged peptides.
Solubility
Miscible with dimethylformamide, hexane, ethyl acetate, chloroform and dichloromethane.
Notes
Incompatible with strong oxidizing agents and water.
Chemical Identifiers
69304-37-6 | |
315.425 | |
DDYAZDRFUVZBMM-UHFFFAOYSA-N | |
172404 | |
CC(C)[Si](C(C)C)(O[Si](C(C)C)(C(C)C)Cl)Cl |
C12H28Cl2OSi2 | |
MFCD00009655 | |
1,3-dichloro-1,1,3,3-tetraisopropyldisiloxane, 1,3-dichlorotetraisopropyldisiloxane, 1,1,3,3-tetraisopropyl-1,3-dichlorodisiloxane, 1,3-dichloro-1,1,3,3-tetraisopropyl-disiloxane, disiloxane, 1,3-dichloro-1,1,3,3-tetrakis 1-methylethyl, 1,1,3,3-tetraisopropyl-1,3-dichlorosiloxane, chloro chlorodiisopropylsilyl oxy diisopropylsilane, chloro-chloro-di propan-2-yl silyl oxy-di propan-2-yl silane, tipdsicl2, tipdsi-cl2Show More | |
chloro-[chloro-di(propan-2-yl)silyl]oxy-di(propan-2-yl)silane |
Specifications
69°C to 70°C (0.5mmHg) | |
C12H28Cl2OSi2 | |
UN1760 | |
>110°C (230°F) | |
Miscible with dimethylformamide,hexane,ethyl acetate,chloroform and dichloromethane. | |
CC(C)[Si](C(C)C)(O[Si](C(C)C)(C(C)C)Cl)Cl | |
315.425 | |
315.43 | |
1.454 | |
0.986 |
69304-37-6 | |
MFCD00009655 | |
1934216 | |
1,3-dichloro-1,1,3,3-tetraisopropyldisiloxane, 1,3-dichlorotetraisopropyldisiloxane, 1,1,3,3-tetraisopropyl-1,3-dichlorodisiloxane, 1,3-dichloro-1,1,3,3-tetraisopropyl-disiloxane, disiloxane, 1,3-dichloro-1,1,3,3-tetrakis 1-methylethyl, 1,1,3,3-tetraisopropyl-1,3-dichlorosiloxane, chloro chlorodiisopropylsilyl oxy diisopropylsilane, chloro-chloro-di propan-2-yl silyl oxy-di propan-2-yl silane, tipdsicl2, tipdsi-cl2 | |
DDYAZDRFUVZBMM-UHFFFAOYSA-N | |
chloro-[chloro-di(propan-2-yl)silyl]oxy-di(propan-2-yl)silane | |
172404 | |
97% | |
Moisture sensitive | |
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P374-P501c
H227-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures