missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromobenzyl)piperazine, 97%, Thermo Scientific™

213.08 SEK - 2425.00 SEK

Chemical Identifiers

CAS 91345-62-9
Molecular Formula C11H15BrN2
Molecular Weight (g/mol) 255.159
MDL Number MFCD02177416
InChI Key MAHWBNAOEVAPJF-UHFFFAOYSA-N
Synonym 1-4-bromobenzyl piperazine, 1-4-bromophenyl methyl piperazine, 1-4-bromo-benzyl-piperazine, piperazine, 1-4-bromophenyl methyl, cbmicro_035222, acmc-20apb8, cambridge id 5862395, 1-4-bromobenzyl-piperazine, 4-bromophenyl methyl piperazine
PubChem CID 876494
IUPAC Name 1-[(4-bromophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=CC=C(C=C2)Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15481298
SDS View Documents
Thermo Scientific Alfa Aesar
H55145.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15461298
SDS View Documents
Thermo Scientific Alfa Aesar
H55145.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15471298
SDS View Documents
Thermo Scientific Alfa Aesar
H55145.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(4-Bromobenzyl)piperazine, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.