missing translation for 'onlineSavingsMsg'
Learn More

1,5-Anhydro-D-sorbitol, 97%, Thermo Scientific Chemicals

2577.59 SEK - 5782.48 SEK

Chemical Identifiers

CAS 154-58-5
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
InChI Key MPCAJMNYNOGXPB-SLPGGIOYSA-N
Synonym 1,5-anhydro-d-glucitol, 1,5-anhydroglucitol, 1,5-anhydrosorbitol, 1,5-anhydro-d-sorbitol, 1-deoxy-d-glucopyranose, aceritol, 1-deoxy-d-glucose, d-glucitol, 1,5-anhydro, unii-54bb3b7xmz, glucitol, 1,5-anhydro
PubChem CID 64960
ChEBI CHEBI:16070
IUPAC Name (2R,3S,4R,5S)-2-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1C(C(C(C(O1)CO)O)O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15559503
View Documents
Thermo Scientific Acros
459912500
250 mg
2577.59 SEK
250mg
Please to purchase this item. Need a web account? Register with us today!
15549503
View Documents
Thermo Scientific Acros
459910010
1 g
5782.48 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

154-58-5
164.16
1,5-anhydro-d-glucitol, 1,5-anhydroglucitol, 1,5-anhydrosorbitol, 1,5-anhydro-d-sorbitol, 1-deoxy-d-glucopyranose, aceritol, 1-deoxy-d-glucose, d-glucitol, 1,5-anhydro, unii-54bb3b7xmz, glucitol, 1,5-anhydro
CHEBI:16070
C1C(C(C(C(O1)CO)O)O)O
C6H12O5
MPCAJMNYNOGXPB-SLPGGIOYSA-N
64960
(2R,3S,4R,5S)-2-(hydroxymethyl)oxane-3,4,5-triol

Specifications

154-58-5
Authentic
1,5-anhydro-d-glucitol, 1,5-anhydroglucitol, 1,5-anhydrosorbitol, 1,5-anhydro-d-sorbitol, 1-deoxy-d-glucopyranose, aceritol, 1-deoxy-d-glucose, d-glucitol, 1,5-anhydro, unii-54bb3b7xmz, glucitol, 1,5-anhydro
C1C(C(C(C(O1)CO)O)O)O
164.16
CHEBI:16070
97%
141.0°C to 144.0°C
C6H12O5
MPCAJMNYNOGXPB-SLPGGIOYSA-N
(2R,3S,4R,5S)-2-(hydroxymethyl)oxane-3,4,5-triol
64960
164.16
1,5-Anhydro-D-sorbitol
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 205-829-3

Product Identifier
  • 1,5-Anhydro-D-sorbitol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.